L-Proline-tert-butyl ester hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: H-Pro-OtBu.HCl

A329666-25G A329666-100G A329666-1G A329666-5G
103825-534EA 5 USD
103825-534 103836-490 76839-460 77277-810 77306-398 77306-400
L-Proline-tert-butyl ester hydrochloride 98%
L-Proline-tert-butyl ester hydrochloride

Caution: for Research Use Only

Formula: C₉H₁₇NO₂
Boiling Pt: 219.2 °C (760 mm Hg)
Melting Pt: 110…112 °C
Flash Pt: 86.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00153459
CAS Number: 5497-76-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR