L(-)-Glutathione (reduced form) 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
L-Glutathione Reduced, GSH, (S)-2-Amino-5-(((R)-1-((carboxymethyl)amino)-3-mercapto-1-oxopropan-2-yl)amino)-5-oxopentanoic acid, Glutathione Reduced
A128528-100G
A128528-10G
A128528-1G
A128528-500G
A128528-25G
A128528-5G
A128528-1000G
103832-030EA
15
USD
103832-030
76797-780
76797-782
76797-784
76797-786
77313-474
77313-476
77666-840
L(-)-Glutathione (reduced form) 97%
L(-)-Glutathione (reduced form)
Caution: for Research Use Only
Formula:
C₁₀H₁₇N₃O₆S Boiling Pt: 754.5 °C (760 mm Hg) Melting Pt: 192…195 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00065939 CAS Number: 70-18-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...