Fmoc-D-Pro-OH, (max. 5% H₂O) 98% (contains < 5% H₂O)

Supplier: Ambeed
Warning

A384263-10G A384263-1G A384263-500G A384263-25G A384263-5G A384263-100G
103837-796EA 6 USD
103837-796 76694-534 76694-536 76694-538 76841-144 76841-146 77551-126
Fmoc-D-Pro-OH, (max. 5% H₂O) 98% (contains < 5% H₂O)
Fmoc-D-Pro-OH

Caution: for Research Use Only

Formula: C₂₀H₁₉NO₄
Boiling Pt: 548.6 °C (760 mm Hg)
Melting Pt: 110…116 °C
Flash Pt: 285.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00077067
CAS Number: 101555-62-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR