exo-Hexahydro-1H-4,7-methanoisoindole-1,3(2H)-dione 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Hexahydro-1H-4,7-methanoisoindole-1,3(2H)-dione, rel-(3aR,4S,7R,7aS)-Hexahydro-1H-4,7-methanoisoindole-1,3(2H)-dione, exo-2,3-Norbornanedicarboximide
A184020-100G
A184020-10G
A184020-1G
A184020-25G
A184020-500G
103815-700EA
5
USD
103815-700
103846-260
77029-560
77029-562
77301-876
77301-878
77871-914
exo-Hexahydro-1H-4,7-methanoisoindole-1,3(2H)-dione 95%
exo-Hexahydro-1H-4,7-methanoisoindole-1,3(2H)-dione
Caution: for Research Use Only
Formula:
C₉H₁₁NO₂ Boiling Pt: 519.7 °C (760 mm Hg) Melting Pt: 173…176 °C Flash Pt: 171 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD12964181 CAS Number: 14805-29-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...