Ethyl-4-amino-2-(methylthio)pyrimidine-5-carboxylate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Ethyl 4-amino-2-(methylthio)pyrimidin-5-carboxylate, Ethyl 4-amino-2-(methylthio)pyrimidine-5-carboxylate
A227698-1G
A227698-25G
A227698-5G
A227698-10G
A227698-250MG
103819-336EA
7
USD
103819-336
103850-244
77089-024
77089-026
77089-028
77288-296
77288-298
Ethyl-4-amino-2-(methylthio)pyrimidine-5-carboxylate 98%
Ethyl 4-amino-2-(methylthio)pyrimidine-5-carboxylate
Caution: for Research Use Only
Formula:
C₈H₁₁N₃O₂S Boiling Pt: 370.3 °C (760 mm Hg) Melting Pt: 133…138 °C Storage Temperature: Ambient |
MDL Number:
MFCD00090781 CAS Number: 776-53-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...