Emtricitabine 98%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-5-fluoro-1-((2R,5S)-2-(hydroxymethyl)-1,3-oxathiolan-5-yl)pyrimidin-2(1H)-one

A188531-1G A188531-100MG A188531-250MG A188531-25G A188531-5G A188531-100G A188531-500G
103816-116EA 9 USD
103816-116 77084-468 77111-674 77111-676 77111-678 77111-680 77593-206 77685-445
Emtricitabine 98%
Emtricitabine

Caution: for Research Use Only

Formula: C₈H₁₀FN₃O₃S
MW: 247.24 g/mol
Boiling Pt: 443.3 °C (760 mm Hg)
Melting Pt: 136…140 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00870151
CAS Number: 143491-57-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR