EDTA tetrasodium salt ≥98% (Loss on drying ≤10%)

Supplier: Ambeed
Danger

A304078-25G A304078-100G
103810-034EA 35 USD
103810-034 77305-894 77626-538
EDTA tetrasodium salt ≥98% (Loss on drying ≤10%)
EDTA tetrasodium salt

Caution: for Research Use Only

Formula: C₁₀H₁₂N₂Na₄O₈
Melting Pt: >300 °C
Flash Pt: 325.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00150027
CAS Number: 64-02-8
ADR: 9,Ⅲ

Order Now

Specification Test Results

Loss on drying ≤ 10%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR