1,12-Dodecanedioic acid 98%
Supplier: Ambeed
|
Warning
|
A101351-100G
A101351-25G
A101351-500G
A101351-1000G
103807-786EA
31
USD
103807-786
103821-610
76809-238
76809-240
76809-242
77297-062
1,12-Dodecanedioic acid 98%
1,12-Dodecanedioic acid
Caution: for Research Use Only
Formula:
C₁₂H₂₂O₄ Boiling Pt: 394 °C (760 mm Hg) Melting Pt: 127…129 °C Flash Pt: 220 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002735 CAS Number: 693-23-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...