DL-Isoleucine 98%

Supplier: Ambeed
Warning

Synonyms: H-DL-Ile-OH

A868688-100G A868688-500G A868688-25G
103841-274EA 18 USD
103841-274 77118-642 77118-644 77308-470
DL-Isoleucine 98%
DL-Isoleucine

Caution: for Research Use Only

Formula: C₆H₁₃NO₂
MW: 131.17 g/mol
Boiling Pt: 225.8 °C (760 mm Hg)
Melting Pt: 275
Storage Temperature: Ambient
MDL Number: MFCD00004268
CAS Number: 443-79-8
Merck Index: 13,05198

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR