Diethyl pyrocarbonate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
DEPC (Diethylpyrocarbonate), Diethyl dicarbonate, DEPC
A127898-10G
A127898-1G
A127898-25G
A127898-5G
A127898-100G
A127898-500G
103831-908EA
8
USD
103831-908
76797-756
76797-758
77298-252
77298-254
77599-860
77624-072
Diethyl pyrocarbonate 97%
Diethyl pyrocarbonate
Caution: for Research Use Only
Formula:
O(COOC₂H₅)₂ Boiling Pt: 93…94 °C at 18 mmHg Flash Pt: 61.3 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00009106 CAS Number: 1609-47-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...