Ciprofloxacin ≥98%

Supplier: Ambeed
Warning

Synonyms: 1-Cyclopropyl-6-fluoro-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid, 1-Cyclopropyl-6-fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl)-3-quinolinecarboxylic Acid

A208061-1000G A208061-100G A208061-500G
103817-850EA 5 USD
103817-850 103848-536 77671-842 77671-844 77883-498
Ciprofloxacin ≥98%
Ciprofloxacin

Caution: Keep in a dark place, inert atmosphere, 2…8 °C

Formula: C₁₇H₁₈FN₃O₃
MW: 331.35 g/mol
Boiling Pt: 581.8 °C (760 mm Hg)
Melting Pt: 255…257 °C (760 mm Hg)
Flash Pt: 305.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00185755
CAS Number: 85721-33-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR