Carbamazepine 98%

Supplier: Ambeed
Warning

Synonyms: 5H-Dibenzo[b,f]azepine-5-carboxamide

A242924-100MG A242924-25MG A242924-25G A242924-100G A242924-1G A242924-500G A242924-5G A242924-5G A242924-10G
77098-518EA 8.49 USD
77098-518 77098-516 77098-514 77098-512 77090-506 77098-522 77098-520 77090-504 103851-316
Carbamazepine 98%
Carbamazepine

Caution: for Research Use Only

Formula: C₁₅H₁₂N₂O
MW: 236.27 g/mol
Boiling Pt: 411 °C (760 mm Hg)
Melting Pt: 189…192 °C
Flash Pt: 202.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00005073
CAS Number: 298-46-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR