N-Boc-L-tyrosine 98%
Supplier: Ambeed
|
Warning
|
A352183-25G
A352183-100G
A352183-10G
A352183-500G
A352183-5G
A352183-1000G
103837-058EA
5
USD
103837-058
76840-172
77278-178
77278-180
77278-182
77306-622
77748-692
N-Boc-L-tyrosine 98%
N-Boc-L-tyrosine
Caution: for Research Use Only
Formula:
C₁₄H₁₉NO₅ Boiling Pt: 484.9 °C (760 mm Hg) Melting Pt: 133…135 °C Flash Pt: 247.1 °C Storage Temperature: Ambient |
MDL Number:
MFCD00037179 CAS Number: 3978-80-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...