Boc-D-Ser-OBzl 95%

Supplier: Ambeed
Warning

Synonyms: N-Boc-D-serine benzyl ester

A151528-250MG A151528-10G A151528-5G A151528-5G A151528-1G A151528-1G A151528-25G
103804-270EA 6 USD
103804-270 77150-376 77150-378 103842-422 77317-998 77317-994 77317-996
Boc-D-Ser-OBzl 95%
Boc-D-Ser-OBzl

Caution: for Research Use Only

Formula: C₁₅H₂₁NO₅
Boiling Pt: 458.8 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD00080265
CAS Number: 141527-78-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR