Azetidine-3-carbonitrile hydrochloride 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
azetidine-3-carbonitrile hydrochloride
A147240-10G
A147240-250MG
A147240-25G
A147240-5G
103803-752EA
23
USD
103803-752
103841-850
76689-228
76689-230
77317-216
77317-218
Azetidine-3-carbonitrile hydrochloride 97%
Azetidine-3-carbonitrile hydrochloride
Caution: for Research Use Only
Formula:
C₄H₇ClN₂ Flash Pt: 91.5 °C Storage Temperature: Freezer |
MDL Number:
MFCD09749828 CAS Number: 345954-83-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...