1-(4-tert-Butylphenyl)-3-(4-methoxyphenyl)-1,3-propanedione 98%

Supplier: Ambeed
Warning

Synonyms: 1-(4-(tert-Butyl)phenyl)-3-(4-methoxyphenyl)propane-1,3-dioneAvobenzone

A325409-5G A325409-25G A325409-10G A325409-5G
76839-282EA 8.49 USD
76839-282 77194-908 103836-346 76839-284
1-(4-tert-Butylphenyl)-3-(4-methoxyphenyl)-1,3-propanedione 98%
1-(4-tert-Butylphenyl)-3-(4-methoxyphenyl)-1,3-propanedione

Caution: for Research Use Only

Formula: C₂₀H₂₂O₃
MW: 310.39 g/mol
Boiling Pt: 463.6 °C (760 mm Hg)
Melting Pt: 81…84 °C
Flash Pt: 203.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00210252
CAS Number: 70356-09-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR