Adenosine-5'-triphosphate disodium salt (ATP disodium salt) 99%

Supplier: Ambeed
Warning

Synonyms: ((2R,3S,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl tetrahydrogen triphosphate, disodium salt, Adenosine 5'-triphosphoric acid disodium, Adenosine 5′-triphosphate disodium salt, ATP 2Na, ATP disodium salt, Adenosine 5ʹ-Triphosphate, Disodium Salt

A197943-100G A197943-10G A197943-1G A197943-5G A197943-1000G A197943-500G
103847-602EA 6 USD
103847-602 76691-110 76691-112 76691-114 77085-616 77625-980 77625-982
Adenosine-5'-triphosphate disodium salt (ATP disodium salt) 99%
Adenosine 5'-triphosphate disodium salt (ATP disodium salt)

Caution: for Research Use Only

Formula: C₁₀H₁₄N₅Na₂O₁₃P₃
MW: 551.14 g/mol
Melting Pt: 175 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00080339
CAS Number: 987-65-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR