Astaxanthine 95%

Supplier: Ambeed
Warning

Synonyms: Carophyll Pink, (3S,3'S)-Astaxanthin, Natupink, (6S,6'S)-3,3'-((1E,3E,5E,7E,9E,11E,13E,15E,17E)-3,7,12,16-Tetramethyloctadeca-1,3,5,7,9,11,13,15,17-nonaene-1,18-diyl)bis(6-hydroxy-2,4,4-trimethylcyclohex-2-enone), BioAstin

A186372-10MG A186372-50MG A186372-1G A186372-5G A186372-10G A186372-25G
103815-932EA 95 USD
103815-932 103846-512 76974-652 76974-654 77302-030 77302-032 77881-398 77881-399
Astaxanthine 95%
Astaxanthine

Caution: for Research Use Only

Formula: C₄₀H₅₂O₄
MW: 596.852 g/mol
Boiling Pt: 774.0±60.0 °C (760 mm Hg)
Melting Pt: 215…216 °C
Flash Pt: 435.8 °C
Storage Temperature: Freezer
MDL Number: MFCD00672621
CAS Number: 472-61-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR