Argatroban monohydrate 98%

Supplier: Ambeed
Warning

Synonyms: MQPA, (2R,4R)-1-[(2S)-5-[(Aminoiminomethyl)amino]-1-oxo-2-[[(1,2,3,4-tetrahydro-3-methyl-8-quinolinyl)sulfonyl]amino]pentyl]-4-methyl-2-piperidinecarboxylic acid, Argipidine, MQPA, (2R,4R)-1-[(2S)-5-[(Aminoiminomethyl)amino]-1-oxo-2-[[(1,2,3,4-tetrahydro-3-methyl-8-quinolinyl)sulfonyl]amino]pentyl]-4-methyl-2-piperidinecarboxylic acid, (2R,4R)-1-[(2S)-5-[(Aminoiminomethyl)amino]-1-oxo-2-[[(1,2,3,4-tetrahydro-3-methyl-8-quinolinyl)sulphonyl]amino]pentyl]-4-methyl-2-piperidinecarboxylic acid, (2R,4R)-1-((2S)-5-Guanidino-2-(3-methyl-1,2,3,4-tetrahydroquinoline-8-sulphonamido)pentanoyl)-4-methylpiperidine-2-carboxylic acid hydrate, Argipidine, (2R,4R)-1-((2S)-5-Guanidino-2-(3-methyl-1,2,3,4-tetrahydroquinoline-8-sulfonamido)pentanoyl)-4-methylpiperidine-2-carboxylic acid hydrate, Argatroban Monohydrate

A115797-1MG A115797-250MG A115797-1G A115797-25MG A115797-50MG A115797-5MG A115797-100MG
103791-434EA 17 USD
103791-434 103792-044 103792-046 103798-624 103799-294 76687-796 76687-798 77311-300 77311-302 77311-304 77311-306 77876-854
Argatroban monohydrate 98%
Argatroban monohydrate

Caution: for Research Use Only

Formula: C₂₃H₃₆N₆O₅S·H₂O
Melting Pt: 176…180 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00895735
CAS Number: 141396-28-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR