2-Anthraceneboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-Anthraceneboronic acid, Anthracen-2-ylboronic acid, 2-Anthraceneboronic Acid

A147103-250MG A147103-1G A147103-5G
103803-738EA 28 USD
103803-738 103841-834 77110-078 77317-192 77317-194
2-Anthraceneboronic acid 98%
2-Anthraceneboronic acid

Caution: for Research Use Only

Formula: C₁₄H₁₁BO₂
MW: 222.05 g/mol
Boiling Pt: 479.5 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD07784001
CAS Number: 141981-64-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR