Amentoflavone 98%

Supplier: Ambeed
Warning

Synonyms: I3′, Didemethyl-ginkgetin, Tridemethylsciadopitysin, II8-Biapigenin, 8-(5-(5,7-Dihydroxy-4-oxo-4H-chromen-2-yl)-2-hydroxyphenyl)-5,7-dihydroxy-2-(4-hydroxyphenyl)-4H-chromen-4-one

A421989-5MG A421989-10MG A421989-25MG A421989-1G A421989-1MG A421989-250MG
103792-708EA 28 USD
103792-708 103792-710 103799-648 76694-966 76842-080 76842-082 77878-171 77878-172 77878-173
Amentoflavone 98%
Amentoflavone

Caution: for Research Use Only

Formula: C₃₀H₁₈O₁₀
MW: 538.464 g/mol
Boiling Pt: 910.5 °C (760 mm Hg)
Melting Pt: 300 °C (760 mm Hg)
Flash Pt: 308.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00017470
CAS Number: 1617-53-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR