Xanthene 98%

Supplier: Ambeed
Warning

Synonyms: 9H-Xanthene, Dibenzopyran

A407918-100G A407918-10G A407918-25G A407918-5G
103838-212EA 17 USD
103838-212 76789-002 76789-004 76841-708 76841-710
Xanthene 98%
Xanthene

Caution: for Research Use Only

Formula: C₁₃H₁₀O
MW: 182.22 g/mol
Boiling Pt: 311 °C (760 mm Hg)
Melting Pt: 100…103 °C (760 mm Hg)
Flash Pt: 137.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00005055
CAS Number: 92-83-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR