6-Nitroindoline 97%

Supplier: Ambeed
Warning

Synonyms: 2,3-Dihydro-6-nitro-(1H)-indole

A719372-1G A719372-5G A719372-25G
103840-730EA 17 USD
103840-730 76844-946 76844-948 77308-192
6-Nitroindoline 97%
6-Nitroindoline

Caution: for Research Use Only

Formula: C₈H₈N₂O₂
MW: 164.16 g/mol
Boiling Pt: 306.6 °C (760 mm Hg)
Melting Pt: 66…70 °C at N/A mmHg
Flash Pt: 139.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005710
CAS Number: 19727-83-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR