6-Bromo-3-chloroisoquinoline 97%
Supplier: Ambeed
|
Warning
|
A416727-1G
A416727-5G
A416727-100MG
A416727-250MG
A416727-10G
103827-324EA
99
USD
103827-324
103838-370
76841-950
76841-952
77033-786
77033-788
77872-661
6-Bromo-3-chloroisoquinoline 97%
6-Bromo-3-chloroisoquinoline
Caution: for Research Use Only
Formula:
C₉H₅BrClN Boiling Pt: 349.5 °C (760 mm Hg) Melting Pt: 100…104 °C Flash Pt: 165.17 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD08703290 CAS Number: 552331-06-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...