6-Aminonicotinaldehyde 96%

Supplier: Ambeed
Warning

Synonyms: 6-aminopyridine-3-carbaldehyde, 6-Aminonicotinaldehyde

A298722-10G A298722-250MG A298722-1G A298722-25G A298722-5G
103814-194EA 15 USD
103814-194 103835-340 76772-580 76772-582 76838-038 76838-040 76838-042
6-Aminonicotinaldehyde 96%
6-Aminonicotinaldehyde

Caution: for Research Use Only

Formula: C₆H₆N₂O
MW: 122.125 g/mol
Melting Pt: 161 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09757481
CAS Number: 69879-22-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR