6-Amino-5-bromoquinoxaline 97%

Supplier: Ambeed
Warning

A230148-100G A230148-10G A230148-1G A230148-25G A230148-5G
103819-526EA 12 USD
103819-526 103850-426 76976-118 76976-120 77089-282 77089-284 77089-286
6-Amino-5-bromoquinoxaline 97%
6-Amino-5-bromoquinoxaline

Caution: for Research Use Only

Formula: C₈H₆BrN₃
Melting Pt: 155…156 °C
Flash Pt: 175.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00799024
CAS Number: 50358-63-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR