5-Phenyl-1H-[1,2,4]triazol-3-ylamine 97%
Supplier: Ambeed
|
Warning
|
A236620-100MG
A236620-250MG
A236620-1G
A236620-5G
103811-114EA
102
USD
103811-114
103850-888
76771-284
76771-286
77089-948
77089-950
5-Phenyl-1H-[1,2,4]triazol-3-ylamine 97%
5-Phenyl-1H-[1,2,4]triazol-3-ylamine
Caution: for Research Use Only
Formula:
C₈H₈N₄ Boiling Pt: 414.4 °C (760 mm Hg) Melting Pt: 191…193 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00086332 CAS Number: 4922-98-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...