5-Methyl-4-nitro-3-isoxazolecarboxylic acid 95%

Supplier: Ambeed
Warning

A101046-250MG A101046-10G A101046-25G
103795-780EA 30 USD
103795-780 103821-470 76994-490 77880-825 77880-826
5-Methyl-4-nitro-3-isoxazolecarboxylic acid 95%
5-Methyl-4-nitro-3-isoxazolecarboxylic acid

Caution: for Research Use Only

Formula: C5H4N2O5
MW: 172.0957 g/mol
Storage Temperature: Freezer
MDL Number: MFCD11520861
CAS Number: 960225-75-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR