5-Bromo-2-fluoropyridine 98%

Supplier: Ambeed
Warning

A117251-100G A117251-10G A117251-25G A117251-500G A117251-5G
103824-730EA 5 USD
103824-730 77271-280 77271-282 77271-284 77271-286 77297-528
5-Bromo-2-fluoropyridine 98%
5-Bromo-2-fluoropyridine

Caution: for Research Use Only

Formula: C₅H₃BrFN
Boiling Pt: 162…164 °C (760 mm Hg)
Flash Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD01863742
CAS Number: 766-11-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR