5-Aminopyridine-2-carbonitrile 96%

Supplier: Ambeed
Danger

Synonyms: 5-Amino-2-pyridinecarbonitrile, 5-Amino-2-cyanopyridine

A217440-1G A217440-25G A217440-5G A217440-100MG A217440-250MG
103818-620EA 22 USD
103818-620 103849-350 77087-970 77087-972 77087-974 77112-646 77112-648
5-Aminopyridine-2-carbonitrile 96%
5-Aminopyridine-2-carbonitrile

Caution: for Research Use Only

Formula: C₆H₅N₃
Boiling Pt: 368.2 °C (760 mm Hg)
Melting Pt: 148…152 °C
Storage Temperature: Ambient
MDL Number: MFCD03093956
CAS Number: 55338-73-3
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR