5-Amino-2-bromobenzoic acid 98%

Supplier: Ambeed
Warning

A273363-25G A273363-5G A273363-10G
103813-138EA 5 USD
103813-138 103853-058 77092-854 77092-856 77166-502
5-Amino-2-bromobenzoic acid 98%
5-Amino-2-bromobenzoic acid

Caution: for Research Use Only

Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
Boiling Pt: 366.2 °C (760 mm Hg)
Flash Pt: 175.2 °C
Storage Temperature: Ambient
MDL Number: MFCD07368968
CAS Number: 2840-02-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR