5-Amino-2-(trifluoromethyl)pyridine 98%

Supplier: Ambeed
Warning

Synonyms: 3-Amino-6-(trifluoromethyl)pyridine, 2-(Trifluoromethyl)pyridin-5-amine, 2-(Trifluoromethyl)-5-pyridylamine

A106917-100G A106917-10G A106917-25G A106917-500G A106917-1G A106917-5G
103797-096EA 14 USD
103797-096 103822-854 77093-780 77093-782 77093-784 77093-786 77283-520 77309-718
5-Amino-2-(trifluoromethyl)pyridine 98%
5-Amino-2-(trifluoromethyl)pyridine

Caution: for Research Use Only

Formula: C₆H₅F₃N₂
Melting Pt: 39…43 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD01862656
CAS Number: 106877-33-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR