4-Cyano-4'-pentylbiphenyl 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4'-Pentyl-4-biphenylcarbonitrile, 4'-Pentyl-[1,1'-biphenyl]-4-carbonitrile
A420052-1G
A420052-25G
A420052-5G
A420052-100G
A420052-10G
A420052-250MG
103827-378EA
6
USD
103827-378
103838-428
76842-048
76842-050
76842-052
77291-400
77291-402
77291-404
4-Cyano-4'-pentylbiphenyl 98%
4-Cyano-4'-pentylbiphenyl
Caution: for Research Use Only
Formula:
C₁₈H₁₉N MW: 249.36 g/mol Boiling Pt: 361.6 °C (760 mm Hg) Melting Pt: 34 °C Flash Pt: 113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00036350 CAS Number: 40817-08-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...