4-Morpholinoaniline 97%
Supplier: Ambeed
|
Warning
|
A293338-100G
A293338-10G
A293338-500G
A293338-1G
A293338-25G
A293338-5G
103813-978EA
5
USD
103813-978
103835-116
76772-468
76772-470
76772-472
76837-728
76837-730
76837-732
4-Morpholinoaniline 97%
4-Morpholinoaniline
Caution: for Research Use Only
Formula:
C₁₀H₁₄N₂O Boiling Pt: 368 °C (760 mm Hg) Melting Pt: 132…135 °C Flash Pt: 176.4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006169 CAS Number: 2524-67-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...