4-Chlorobenzenesulfonyl chloride 96%

Supplier: Ambeed
Danger

A237135-100G A237135-500G A237135-25G A237135-5G
103850-910EA 5 USD
103850-910 76826-644 76826-646 76826-648 77089-980 77304-074
4-Chlorobenzenesulfonyl chloride 96%
4-Chlorobenzenesulphonyl chloride

Caution: for Research Use Only

Formula: C₆H₄Cl₂O₂S
Boiling Pt: 141 °C (760 mm Hg)
Melting Pt: 50…52 °C
Flash Pt: 108 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007439
CAS Number: 98-60-2
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR