4-Chlorobenzaldehyde 98%
Supplier: Ambeed
|
Warning
|
A300364-100G
A300364-25G
A300364-500G
103810-016EA
48
USD
103810-016
76977-712
76977-714
76977-716
4-Chlorobenzaldehyde 98%
4-Chlorobenzaldehyde
Caution: for Research Use Only
Formula:
C₇H₅ClO Boiling Pt: 213.7 °C (760 mm Hg) Melting Pt: 46 °C Flash Pt: 87 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003379 CAS Number: 104-88-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...