4-Bromobenzamidine hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: 4-Bromophenylamidine hydrochloride, 4-Bromobenzimidamide hydrochloride, 4-Bromobenzene-1-carboximidamide hydrochloride

A124216-100G A124216-10G A124216-250MG A124216-25G A124216-1G A124216-5G
103800-506EA 13 USD
103800-506 103831-346 77161-500 77161-502 77161-504 77161-506 77297-978 77312-640
4-Bromobenzamidine hydrochloride 98%
4-Bromobenzamidine hydrochloride

Caution: for Research Use Only

Formula: C₇H₈BrClN₂
Storage Temperature: Ambient
MDL Number: MFCD00464967
CAS Number: 55368-42-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR