4-Bromo-3-hydroxypyridine 98%
Supplier: Ambeed
|
Warning
|
A149386-100MG
A149386-10G
A149386-250MG
A149386-1G
A149386-5G
103804-038EA
14
USD
103804-038
103842-156
76997-280
76997-282
76997-284
77317-648
77317-650
4-Bromo-3-hydroxypyridine 98%
4-Bromo-3-hydroxypyridine
Caution: for Research Use Only
Formula:
C₅H₄BrNO Boiling Pt: 316.2 °C (760 mm Hg) Melting Pt: 134…136 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03093021 CAS Number: 161417-28-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...