4-Bromo-2-chlorobenzonitrile 98%
Supplier: Ambeed
|
Danger
|
A301888-5G
A301888-100G
A301888-10G
A301888-500G
A301888-1G
A301888-25G
103814-314EA
5
USD
103814-314
103835-470
76838-164
77002-532
77002-534
77002-536
77305-836
77305-838
4-Bromo-2-chlorobenzonitrile 98%
4-Bromo-2-chlorobenzonitrile
Caution: for Research Use Only
Formula:
C₇H₃BrClN Boiling Pt: 279.8 °C (760 mm Hg) Melting Pt: 67…68 °C Storage Temperature: Ambient |
MDL Number:
MFCD00040883 CAS Number: 154607-01-9 UN: 3439 ADR: 6.1,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...