4-Bromo-2,6-dichloropyridine 98%
Supplier: Ambeed
|
Warning
|
A179633-10G
A179633-250MG
A179633-1G
A179633-25G
A179633-5G
103807-448EA
8
USD
103807-448
103845-800
76813-512
76813-514
77083-268
77083-270
77083-272
4-Bromo-2,6-dichloropyridine 98%
4-Bromo-2,6-dichloropyridine
Caution: for Research Use Only
Formula:
C₅H₂BrCl₂N Boiling Pt: 254.3 °C (760 mm Hg) Melting Pt: 95 °C Flash Pt: 107.597 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD11848646 CAS Number: 98027-80-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...