4-Aminoquinoline 95%
Supplier: Ambeed
|
Warning
|
A224551-25G
A224551-5G
A224551-10G
A224551-250MG
A224551-1G
103819-120EA
16
USD
103819-120
103850-012
77088-672
77088-674
77165-318
77165-320
77303-702
4-Aminoquinoline 95%
4-Aminoquinoline
Caution: for Research Use Only
Formula:
C₉H₈N₂ Boiling Pt: 332.3 °C (760 mm Hg) Melting Pt: 154…155 °C Flash Pt: 181.2 °C Storage Temperature: Ambient |
MDL Number:
MFCD00463448 CAS Number: 578-68-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...