4-Amino-3,5-difluorobenzonitrile 98%

Supplier: Ambeed
Warning

A187672-10G A187672-250MG A187672-1G A187672-25G A187672-5G
103816-034EA 13 USD
103816-034 103846-624 76974-708 76974-710 77084-328 77084-330 77084-332
4-Amino-3,5-difluorobenzonitrile 98%
4-Amino-3,5-difluorobenzonitrile

Caution: for Research Use Only

Formula: C₇H₄F₂N₂
MW: 154.1169 g/mol
Melting Pt: 110…111 °C
Storage Temperature: Ambient
MDL Number: MFCD03407467
CAS Number: 110301-23-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR