4-[[(4-Fluorophenyl)imino]methyl]phenol 98%

Supplier: Ambeed
Danger

Synonyms: ALPHA-(4-FLUOROPHENYLIMINO)-P-CRESOL, 4-[[(4-Fluorophenyl)imino]methyl]phenol

A240874-1G A240874-5G
103811-370EA 6 USD
103811-370 103851-176 77090-332 77090-334
4-[[(4-Fluorophenyl)imino]methyl]phenol 98%
4-[[(4-Fluorophenyl)imino]methyl]phenol

Caution: for Research Use Only

Formula: C₁₃H₁₀FNO
Boiling Pt: 370.9 °C (760 mm Hg) (lit.)
Melting Pt: 183.0 °C (760 mm Hg) (lit.)
Flash Pt: 178 °C (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00029739
CAS Number: 3382-63-6
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR