4,4,7a-Trimethyl-5,6,7,7a-tetrahydrobenzofuran-2(4H)-one 97%

Supplier: Ambeed
Warning

A108106-100G A108106-100MG A108106-10G A108106-250MG A108106-25G A108106-1G A108106-5G
103797-298EA 14 USD
103797-298 103823-074 76765-620 76765-622 76765-624 76765-626 76765-628 77283-692 77309-920
4,4,7a-Trimethyl-5,6,7,7a-tetrahydrobenzofuran-2(4H)-one 97%
4,4,7a-Trimethyl-5,6,7,7a-tetrahydrobenzofuran-2(4H)-one

Caution: for Research Use Only

Formula: C₁₁H₁₆O₂
MW: 180.2435 g/mol
Flash Pt: 120.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06409997
CAS Number: 15356-74-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR