4,4,5,5-Tetramethyl-2-(oxetan-3-yl)-1,3,2-dioxaborolane 97%

Supplier: Ambeed
Warning

A117098-100MG A117098-250MG A117098-1G
103798-828EA 162 USD
103798-828 103824-702 77161-062 77161-064 77311-488
4,4,5,5-Tetramethyl-2-(oxetan-3-yl)-1,3,2-dioxaborolane 97%
4,4,5,5-Tetramethyl-2-(oxetan-3-yl)-1,3,2-dioxaborolane

Caution: for Research Use Only

Formula: C₉H₁₇BO₃
Storage Temperature: Freezer
MDL Number: MFCD26521383
CAS Number: 1396215-84-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR