2-Amino-4-trifluoromethyl-1,3-thiazole 97%

Supplier: Ambeed
Warning

Synonyms: 4-(trifluoromethyl)thiazol-2-amine, 2-Amino-4-trifluoromethyl-1,3-thiazole, 4-(trifluoromethyl)-1,3-thiazol-2-amine, 4-Trifluoromethyl-thiazole-2ylamine, 2-Amino-4-trifluoromethylthiazole, 4-(Trifluoromethyl)thiazol-2-amine

A159971-10G A159971-250MG A159971-1G A159971-5G
103805-308EA 10 USD
103805-308 103843-536 76812-670 76812-672 77319-442 77319-444
2-Amino-4-trifluoromethyl-1,3-thiazole 97%
2-Amino-4-trifluoromethyl-1,3-thiazole

Caution: for Research Use Only

Formula: C₄H₃F₃N₂S
MW: 168.14 g/mol
Boiling Pt: 214 °C (760 mm Hg)
Melting Pt: 60…62 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00832759
CAS Number: 349-49-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR