4-(4-Amino-3-fluorophenoxy)-N-methyl-2-pyridinecarboxamide 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-(4-Amino-3-fluorophenoxy)-N-methylpicolinamide
A353233-1G
A353233-25G
A353233-5G
A353233-250MG
A353233-100G
103826-090EA
53
USD
103826-090
103837-092
76840-208
76840-210
76840-212
76989-680
77885-930
4-(4-Amino-3-fluorophenoxy)-N-methyl-2-pyridinecarboxamide 97%
4-(4-Amino-3-fluorophenoxy)-N-methyl-2-pyridinecarboxamide
Caution: for Research Use Only
Formula:
C₁₃H₁₂FN₃O₂ Flash Pt: 231.9 °C Storage Temperature: Ambient |
MDL Number:
MFCD09033852 CAS Number: 757251-39-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...