3-Iodooxetane 98%

Supplier: Ambeed
Danger

Synonyms: 2-Iodo-1,3-epoxypropane, 1,3-Epoxy-2-iodopropane

A240486-25G A240486-5G A240486-10G A240486-1G
103811-350EA 5 USD
103811-350 103851-152 77090-304 77090-306 77098-462 77304-194
3-Iodooxetane 98%
3-Iodooxetane

Caution: for Research Use Only

Formula: C₃H₅IO
MW: 183.98 g/mol
Boiling Pt: 165.9 °C (760 mm Hg)
Flash Pt: 54 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08544403
CAS Number: 26272-85-5
UN: 1993
ADR: 3 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR