3-Iodobenzoic acid 98%

Supplier: Ambeed
Warning

Synonyms: m-Iodobenzoic acid

A246279-100G A246279-10G A246279-500G A246279-1000G A246279-25G
103809-756EA 293 USD
103809-756 103851-532 76771-544 76771-546 76771-548 77304-382 77304-384
3-Iodobenzoic acid 98%
3-Iodobenzoic acid

Caution: for Research Use Only

Formula: C₇H₅IO₂
Boiling Pt: 337.2 °C (760 mm Hg)
Melting Pt: 185…189 °C
Flash Pt: 157.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00002496
CAS Number: 618-51-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR