3-(Dimethylamino)phenol 97%

Supplier: Ambeed
Warning

A442672-25G A442672-100G A442672-10G A442672-5G
103838-760EA 7 USD
103838-760 76842-532 77279-136 77279-138 77307-320
3-(Dimethylamino)phenol 97%
3-(Dimethylamino)phenol

Caution: for Research Use Only

Formula: C₈H₁₁NO
Boiling Pt: 267 °C (760 mm Hg)
Melting Pt: 80…85 °C
Flash Pt: 148 °C
Storage Temperature: Ambient
MDL Number: MFCD00002264
CAS Number: 99-07-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR